Borane-dimethylamine complex, 98+%
Borane-dimethylamine complex, 98+%
Borane-dimethylamine complex, 98+%
Borane-dimethylamine complex, 98+%
Thermo Scientific Chemicals

Borane-dimethylamine complex, 98+%

CAS: 74-94-2 | C2H10BN | 58.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 177315000
Price (USD)
813.65
Online Exclusive
904.00
Save 90.35 (10%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
813.65
Online Exclusive
904.00
Save 90.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS74-94-2
IUPAC Namedimethylamine borane
Molecular FormulaC2H10BN
InChI KeyFTDUHBOCJSQEKS-UHFFFAOYSA-N
SMILESB.CNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules or crystals
Melting point33°C to 37°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with Na2S2O3>=98.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473928Certificate of AnalysisJun 27, 2025177315000, 177310050, 177310250, 177311000
A0468116Certificate of AnalysisJan 31, 2025177315000, 177310050, 177310250, 177311000
A0443740Certificate of AnalysisJan 31, 2025177315000, 177310050, 177310250, 177311000
A0469534Certificate of AnalysisJan 29, 2025177315000, 177310050, 177310250, 177311000
A0465709Certificate of AnalysisAug 26, 2024177315000, 177310050, 177310250, 177311000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection