Thermo Scientific Chemicals

1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 177481000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 177481000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 177481000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2579-20-6
IUPAC Name
[(1R,3S)-3-(azaniumylmethyl)cyclohexyl]methanaminium
Molecular Formula
C8H20N2
InChI Key
QLBRROYTTDFLDX-OCAPTIKFSA-P
SMILES
[NH3+]C[C@H]1CCC[C@@H](C[NH3+])C1
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
GC
>=98.5 %
Appearance (Form)
Liquid
Refractive index
1.4920 to 1.4940 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text