trans,trans-2,4-Decadienal, 95%, Tech., Thermo Scientific Chemicals
trans,trans-2,4-Decadienal, 95%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans,trans-2,4-Decadienal, 95%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 177510010
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.70 
Save 4.05 (14%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.70 
Save 4.05 (14%)
Add to cart
trans,trans-2,4-Decadienal, 95%, Tech., Thermo Scientific Chemicals
Catalog number177510010
Price (USD)/ Each
25.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.70 
Save 4.05 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2E,4E)-deca-2,4-dienal
Molecular FormulaC10H16O
InChI KeyJZQKTMZYLHNFPL-BLHCBFLLSA-N
SMILESCCCCC\C=C\C=C\C=O
Molecular Weight (g/mol)152.24
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=92 % (main isomer)
Refractive index1.5140 to 1.5180 (20°C, 589 nm)
GC>=94 % (sum of isomers)
Appearance (Color)Clear yellow
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only