Thermo Scientific Chemicals

Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific Chemicals

Catalog number: 177610250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific Chemicals

Catalog number: 177610250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 177610250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4202-14-6
IUPAC Name
dimethyl (2-oxopropyl)phosphonate
Molecular Formula
C5H11O4P
InChI Key
UOWIYNWMROWVDG-UHFFFAOYSA-N
SMILES
COP(=O)(CC(C)=O)OC
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.4380 to 1.4410 (20°C, 589 nm)
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text