Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Thermo Scientific Chemicals

Cellulose acetate, average M.W. 100,000

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Catalog number 177780250
Price (USD)/ Each
18.65
Online exclusive
20.90 
Save 2.25 (11%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
18.65
Online exclusive
20.90 
Save 2.25 (11%)
Add to cart
Cellulose acetate, average M.W. 100,000
Catalog number177780250
Price (USD)/ Each
18.65
Online exclusive
20.90 
Save 2.25 (11%)
-
Add to cart
Chemical Identifiers
CAS9004-35-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Acetyl content39.8 % (typical)
Hydroxyl content3.6 % (typical)
Loss on drying=<5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only