Thermo Scientific Chemicals

Poly(acrylamide), granular, non-ionic, ∽ M.W. 5 to 6.000.000, Thermo Scientific Chemicals

Catalog number: 178042500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Poly(acrylamide), granular, non-ionic, ∽ M.W. 5 to 6.000.000, Thermo Scientific Chemicals

Catalog number: 178042500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 178042500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-05-8
Molecular Formula
(C3H5NO)n
SMILES
NC(=O)C(-*)C-*
MDL Number
MFCD00084392
Loss on drying
=<13 %
Appearance (Color)
White to light yellow
Appearance (Form)
Granules
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text