Accucore™ Biphenyl Reversed Phase HPLC Columns
Accucore™ Biphenyl Reversed Phase HPLC Columns
Accucore™ Biphenyl Reversed Phase HPLC Columns
Accucore™ Biphenyl Reversed Phase HPLC Columns
Accucore™ Biphenyl Reversed Phase HPLC Columns
Thermo Scientific™

Accucore™ Biphenyl Reversed Phase HPLC Columns

Use this solid core column for fast and reliable separation of critical pairs and isomers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
17826-01210510 mm
17826-05213050 mm
17826-102130100 mm
Catalog number 17826-012105
Price (USD)
604.83
Pack of 4
Add to cart
Length (Metric):
10 mm
Price (USD)
604.83
Pack of 4
Add to cart
Ask our AI about this Product

Choose Thermo Scientific™ Accucore™ Biphenyl LC Columns for fast and reliable separation of critical pairs and isomers. These columns provide unique selectivity for aromatic and moderately polar analytes. Their optimized packing offers a rugged platform for a variety of matrices. Based on Core Enhanced Technology, Accucore liquid chromatography reversed phase columns provide fast, high-resolution separations without the elevated backpressures seen using sub-2μm particles.

  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Mixed-mode selectivity for aromatic and moderately polar analytes
  • Enhanced Pi-pi interactions with aromatics
  • Excellent lot-to-lot reproducibility

Trust your separation of critical pairs and isomers to Accucore Biphenyl LC Columns. These columns offer fast run times, excellent robustness, and consistent reproducibility, and provide the perfect balance of polar analyte retention and hydrophobic analyte elution.

Specifications
Column FormatGuard Cartridge
Column TypeReversed Phase
EndcappedYes
Flow Rate0.4 mL/min
For Use WithHPLC/UHPLC Instrumentation
Max. Pressure8700 psi (600 bar)
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Particle ShapeSpherical Solid Core
Stationary PhaseBiphenyl
Surface Area130 m²/g
Temperature70°C
USP TypeL11
Carbon Load6%
Diameter (Metric)2.1 mm
Length (Metric)10 mm
Particle Size2.6 μm
PhaseReversed Phase
Pore Size80 Å
Product LineAccucore
pH2 to 8
Unit SizePack of 4
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Share catalog number, name or link

1x1 image pixel for data collection