Polycaprolactone, M.W. approx. 80,000
Polycaprolactone, M.W. approx. 80,000
Thermo Scientific Chemicals

Polycaprolactone, M.W. approx. 80,000

CAS: 24980-41-4 | (C6H10O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 178305000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS24980-41-4
Molecular Formula(C6H10O2)n
SMILES*-CCCCCC(=O)O-*
MDL NumberMFCD00084404
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Beads
Infrared spectrumConforms
Water=<0.5 %
Molecular weight80000 to 85000 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link