Polycarbonate resin, ∽ M.W. 45.000, pellets, Thermo Scientific Chemicals
Polycarbonate resin, ∽ M.W. 45.000, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polycarbonate resin, ∽ M.W. 45.000, pellets, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
Packaging:
Glass bottle
Catalog number 178315000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24936-68-3
Molecular Formula(C16H14O3)n
SMILESCC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1
MDL NumberMFCD00084476
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Appearance (Form)Pellets
Infrared spectrumConforms
Loss on drying=<2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only