Thermo Scientific Chemicals

Poly(dimethylsiloxane), Thermo Scientific Chemicals

Catalog number: 178445000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Poly(dimethylsiloxane), Thermo Scientific Chemicals

Catalog number: 178445000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 178445000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9016-00-6
Molecular Formula
(C2H6OSi)n
SMILES
C[Si](C)(-*)O-*
MDL Number
MFCD00084411
Appearance (Form)
Clear viscous liquid
Viscosity
95 to 105 cSt (at 25°C)
Refractive index
1.4020 to 1.4050 (20°C, 589 nm)
Volatiles
=<0.5 % (200°C)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text