Poly(ethylene oxide), M.W. 250,000 to 400,000
Poly(ethylene oxide), M.W. 250,000 to 400,000
Poly(ethylene oxide), M.W. 250,000 to 400,000
Poly(ethylene oxide), M.W. 250,000 to 400,000
Thermo Scientific Chemicals

Poly(ethylene oxide), M.W. 250,000 to 400,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 178602500
Price (USD)
86.65
Special offer
Online exclusive
102.00
Save 15.35 (15%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
102.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Loss on drying=<2 % (10 g, 50°C, 5 h, 10 mmHg)
Infrared spectrumConforms
Appearance (Form)Powder
Viscosity2000 to 4000 mPa.s (10 % w/w in water at 25°C)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469378Certificate of AnalysisJan 29, 2025178602500, 178600010
A0441858Certificate of AnalysisFeb 27, 2023178602500, 178600010
A0398604Certificate of AnalysisJan 14, 2021178602500, 178600010
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection