Thermo Scientific Chemicals

Poly(methyl methacrylate), beads, average M.W. 35,000, Thermo Scientific Chemicals

Catalog number: 178760010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Poly(methyl methacrylate), beads, average M.W. 35,000, Thermo Scientific Chemicals

Catalog number: 178760010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 178760010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9011-14-7
Molecular Formula
(C5H8O2)n
SMILES
COC(=O)C(C)(-*)C-*
MDL Number
MFCD00134349
Appearance (Color)
White
Appearance (Form)
Beads
Infrared spectrum
Conforms
Loss on drying
=<5 %
Particle size
140 to 250 microns (approximately)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text