Poly(vinyl alcohol), 98.0-98.8% hydrolyzed, M.W. approx. 31,000-50,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0-98.8% hydrolyzed, M.W. approx. 31,000-50,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinyl alcohol), 98.0-98.8% hydrolyzed, M.W. approx. 31,000-50,000, Thermo Scientific Chemicals

Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Catalog number 178971000
Price (USD)/ Each
23.65
Online exclusive
26.50 
Save 2.85 (11%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
23.65
Online exclusive
26.50 
Save 2.85 (11%)
Add to cart
Poly(vinyl alcohol), 98.0-98.8% hydrolyzed, M.W. approx. 31,000-50,000, Thermo Scientific Chemicals
Catalog number178971000
Price (USD)/ Each
23.65
Online exclusive
26.50 
Save 2.85 (11%)
-
Add to cart
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Volatile matter=<5.0 %
Infrared spectrumConforms
Viscosity5.5 to 6.6 cP (4 % in water)
pH5.0 to 7.0 (4 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only