Thermo Scientific Pierce Phenol-Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.
Features of Phenol:Chloroform + Tris Buffer :
• Offers excellent, nuclease-free performance • Phenol is supplied premixed with Chloroform:Isoamyl Alcohol (25:24:1) • Supplied at pH 6.7 for special, lower pH DNA extractions; for most DNA applications, the alkaline buffer (included) should be added to produce a pH of 8.0
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)
DNA Extraction
Product Type
Phenol Chloroform and Tris Buffer
Quantity
400 mL
Form
Liquid
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.