2-(Benzyloxy)phenol, 97%, Thermo Scientific Chemicals
2-(Benzyloxy)phenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Benzyloxy)phenol, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number 179320050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6272-38-4
IUPAC Name2-(benzyloxy)phenol
Molecular FormulaC13H12O2
InChI KeyCCZCXFHJMKINPE-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
GC>=95.0 %
Appearance (Color)Colorless to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only