6-Methoxyquinoline, 98%, Thermo Scientific Chemicals
6-Methoxyquinoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methoxyquinoline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 179341000
Price (USD)/ Each
180.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
212.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
180.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
212.00 
Save 31.35 (15%)
Add to cart
6-Methoxyquinoline, 98%, Thermo Scientific Chemicals
Catalog number179341000
Price (USD)/ Each
180.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
212.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
CAS5263-87-6
IUPAC Name6-methoxyquinoline
Molecular FormulaC10H9NO
InChI KeyHFDLDPJYCIEXJP-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
Refractive index1.6220 to 1.6260 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear yellow to pink to purple to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only