Tetrathiafulvalene, 99+%, Thermo Scientific Chemicals
Tetrathiafulvalene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrathiafulvalene, 99+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 mg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 179412500
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.10 
Save 9.45 (14%)
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.10 
Save 9.45 (14%)
Add to cart
Tetrathiafulvalene, 99+%, Thermo Scientific Chemicals
Catalog number179412500
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.10 
Save 9.45 (14%)
-
Add to cart
Chemical Identifiers
CAS31366-25-3
IUPAC Name2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole
Molecular FormulaC6H4S4
InChI KeyFHCPAXDKURNIOZ-UHFFFAOYSA-N
SMILESS1C=CSC1=C1SC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point117°C to 120°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only