Thermo Scientific Chemicals

Phenylselenyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 179510250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenylselenyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 179510250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 179510250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5707-04-0
IUPAC Name
(phenylselanyl)chlorane
Molecular Formula
C6H5ClSe
InChI Key
WJCXADMLESSGRI-UHFFFAOYSA-N
SMILES
Cl[Se]C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
Orange to dark red
Melting point
59°C to 64°C (Dec)
Appearance (Form)
Crystalline powder or crystals and/or chunks
Titration Argentometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text