4-Nitrophenethyl alcohol, 99%, Thermo Scientific Chemicals
4-Nitrophenethyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenethyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 179610100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(4-nitrophenyl)ethan-1-ol
Molecular FormulaC8H9NO3
InChI KeyIKMXRUOZUUKSON-UHFFFAOYSA-N
SMILESOCCC1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol)167.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
HPLC>=98.5 %
Appearance (Form)Powder
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only