Nunc™ Cell Scrapers
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Cell Scrapers
Thermo Scientific™

Nunc™ Cell Scrapers

Conduct a variety of applications with sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scrapers, which feature an adjustable blade.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric) Handle
17969323 cm
179693PK23 cm
17970732 cm
179707PK32 cm
Catalog number 179693
Price (USD)
1,071.65
Online Exclusive
1,170.00
Save 98.35 (8%)
Case of 250
Estimated availability date 06-Aug-2025
Add to cart
Length (Metric) Handle:
23 cm
Price (USD)
1,071.65
Online Exclusive
1,170.00
Save 98.35 (8%)
Case of 250
Add to cart
Featuring an adjustable blade, the sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scraper allows for convenient mechanical harvesting of the adherent cells from culture vessels. Comes in 2 lengths to accommodate wide range of culture wares.
Features:
  • Flexible polystyrene handles
  • Sterilized and individually wrapped

Ordering Information:

  • 179707: for use with 75 to 175cm2 flasks
  • 179693: for use with 25 to 80cm2 flasks
Warranty: 90 days.
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)25 to 80 cm&Sup2; Flask
Length (English) Handle9 in.
Length (Metric) Handle23 cm
No. per Case250/Cs.
No. per Pack1/Pk.
SterilitySterile
Length (English) Blade0.2952 in.
Length (Metric) Blade7.5 mm
MaterialPolystyrene (Handle)
Product TypeCell Scraper
Width (English) Blade0.6889 in.
Width (Metric) Blade17.5 mm
Unit SizeCase of 250

Frequently asked questions (FAQs)

Can I use a Nunc Cell Scraper with a Nunc TripleFlask that has Nunclon Delta surface treatment?

The design of the Nunc TripleFlask does not allow for mechanical scraping. However, cells are easily harvested using protein digesting enzymes, such as trypsin, collagenase, and Pronase or by using non-enzymatic dissociation solutions.
The following is a document link regarding culturing techniques for Nunc TripleFlask:
https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/culturing-technique-nunc-tripleflask-technical-note.pdf

Are your Nunc Cell Scrapers flexible or rigid? What material are they made of?

Our Nunc Cell Scraper blade is rigid and made from polyethylene. It is hinged to allow for positioning at different angles for scraping cells using the windshield wiper or hoe method.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection