Thermo Scientific Chemicals

2-Iodobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 179700100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Iodobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 179700100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 179700100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5159-41-1
IUPAC Name
(2-iodophenyl)methanol
Molecular Formula
C7H7IO
InChI Key
WZCXOBMFBKSSFA-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=C1I
Appearance (Color)
White to yellow to pink
Appearance (Form)
Powder or needles
Infrared spectrum
Conforms
Melting point
88°C to 93°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text