2-bromonaphtalène, 99 %, Thermo Scientific Chemicals
2-bromonaphtalène, 99 %, Thermo Scientific Chemicals
2-bromonaphtalène, 99 %, Thermo Scientific Chemicals
2-bromonaphtalène, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-bromonaphtalène, 99 %, Thermo Scientific Chemicals

CAS: 580-13-2 | C10H7Br | 207.07 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
100 g
5 g
25 g
Conditionnement:
Flacon en verre
Référence 179711000
Prix (EUR)/ Each
253,30
Offre exceptionnelle en ligne
298,00
Économisez 44,70 (15%)
Ajouter au panier
Quantité:
100 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)/ Each
253,30
Offre exceptionnelle en ligne
298,00
Économisez 44,70 (15%)
Ajouter au panier
Identifiants chimiques
CAS580-13-2
Appearance (Color)White to light yellow to light beige
Appearance (Form)Crystalline powder and/or chunks
GC>=98.5 %
Infrared spectrumConforms
Melting point53°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Partager la référence catalogue, le nom ou le lien.