Thermo Scientific Chemicals

3,3-Dimethylacryloyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 179730250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,3-Dimethylacryloyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 179730250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 179730250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3350-78-5
IUPAC Name
3-methylbut-2-enoyl chloride
Molecular Formula
C5H7ClO
InChI Key
BDUBTLFQHNYXPC-UHFFFAOYSA-N
SMILES
CC(C)=CC(Cl)=O
GC
>=96.0 %
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Appearance
May darken on storage
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text