Thermo Scientific Chemicals

2'-Bromoacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: 179760250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2'-Bromoacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: 179760250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 179760250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2142-69-0
IUPAC Name
1-(2-bromophenyl)ethan-1-one
Molecular Formula
C8H7BrO
InChI Key
PIMNFNXBTGPCIL-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=C1Br
Appearance (Color)
Clear light yellow to orange
Refractive index
1.5650 to 1.5690 (20°C, 589 nm)
GC
>=98.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text