Thermo Scientific Chemicals

2'-Fluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: 179780250
Each
Thermo Scientific Chemicals

2'-Fluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: 179780250
Each
Catalog number: 179780250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
445-27-2
IUPAC Name
1-(2-fluorophenyl)ethan-1-one
Molecular Formula
C8H7FO
InChI Key
QMATYTFXDIWACW-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=C1F
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow
GC
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.5065 to 1.5085 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text