Thermo Scientific Chemicals

4-chlorosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 179840250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-chlorosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 179840250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 179840250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5106-98-9
IUPAC Name
4-chloro-2-hydroxybenzoic acid
Molecular Formula
C7H5ClO3
InChI Key
LWXFCZXRFBUOOR-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Cl)C=C1O
Appearance (Color)
Off-white to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
209°C to 213°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text