Methyl 2-chloroacetoacetate, 95%, Thermo Scientific Chemicals
Methyl 2-chloroacetoacetate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-chloroacetoacetate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 179900100
Price (USD)/ Each
88.80
-
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
88.80
Add to cart
Methyl 2-chloroacetoacetate, 95%, Thermo Scientific Chemicals
Catalog number179900100
Price (USD)/ Each
88.80
-
Add to cart
Chemical Identifiers
CAS4755-81-1
IUPAC Namemethyl 2-chloro-3-oxobutanoate
Molecular FormulaC5H7ClO3
InChI KeyGYQRIAVRKLRQKP-UHFFFAOYNA-N
SMILESCOC(=O)C(Cl)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Clear yellow
Refractive index1.4455 to 1.4475 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only