Thermo Scientific Chemicals

Methyl 2-chloroacetoacetate, 95%, Thermo Scientific Chemicals

Catalog number: 179900500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 2-chloroacetoacetate, 95%, Thermo Scientific Chemicals

Catalog number: 179900500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 179900500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4755-81-1
IUPAC Name
methyl 2-chloro-3-oxobutanoate
Molecular Formula
C5H7ClO3
InChI Key
GYQRIAVRKLRQKP-UHFFFAOYNA-N
SMILES
COC(=O)C(Cl)C(C)=O
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=94 %
Appearance (Color)
Clear yellow
Refractive index
1.4455 to 1.4475 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text