p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 179930010
Price (USD)/ Each
435.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
512.00 
Save 76.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
435.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
512.00 
Save 76.35 (15%)
Add to cart
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
Catalog number179930010
Price (USD)/ Each
435.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
512.00 
Save 76.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,4-dithiane-2,5-diol
Molecular FormulaC4H8O2S2
InChI KeyYUIOPHXTILULQC-UHFFFAOYNA-N
SMILESOC1CSC(O)CS1
Molecular Weight (g/mol)152.23
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95.0 %
Appearance (Color)White to yellow
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only