Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals
Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 180070100
Price (USD)/ Each
40.10
-
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
40.10
Add to cart
Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals
Catalog number180070100
Price (USD)/ Each
40.10
-
Add to cart
Chemical Identifiers
CAS32001-55-1
IUPAC Namehydrogen pyridine fluoride
Molecular FormulaC5H6FN
InChI KeyGRJJQCWNZGRKAU-UHFFFAOYSA-N
SMILES[H+].[F-].C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH65 to 70 % (HF)
Appearance (Form)Liquid
Appearance (Color)Clear light yellow to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only