Thermo Scientific Chemicals

Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals

Catalog number: 180070500
50 g, Plastic Bottle in Tin Can, Each
Thermo Scientific Chemicals

Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals

Catalog number: 180070500
50 g, Plastic Bottle in Tin Can, Each
Quantity
Packaging
Catalog number: 180070500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32001-55-1
IUPAC Name
hydrogen pyridine fluoride
Molecular Formula
C5H6FN
InChI Key
GRJJQCWNZGRKAU-UHFFFAOYSA-N
SMILES
[H+].[F-].C1=CC=NC=C1
Titration with NaOH
65 to 70 % (HF)
Appearance (Form)
Liquid
Appearance (Color)
Clear light yellow to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text