Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine
Thermo Scientific Chemicals

Hydrogen fluoride-pyridine, approx. 70% HF, approx. 30% Pyridine

CAS: 32001-55-1 | C5H6FN | 99.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
50 g
Packaging:
View all
Plastic Bottle in Tin Can
Catalog number 180071000
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle in Tin Can
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS32001-55-1
IUPAC Namehydrogen pyridine fluoride
Molecular FormulaC5H6FN
InChI KeyGRJJQCWNZGRKAU-UHFFFAOYSA-N
SMILES[H+].[F-].C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration with NaOH65 to 70 % (HF)
Appearance (Color)Clear light yellow to light brown
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0446017Certificate of AnalysisApr 04, 2024180071000, 180070500, 180070100
A0433235Certificate of AnalysisMay 02, 2023180070100, 180070500, 180071000
A0395525Certificate of AnalysisDec 15, 2022180071000, 180070500, 180070100
A0393770Certificate of AnalysisDec 15, 2022180071000, 180070500, 180070100
A0411133Certificate of AnalysisDec 13, 2022180071000, 180070500, 180070100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link