DNA Polymerase I
DNA Polymerase I
Invitrogen™

DNA Polymerase I

DNA Polymerase I is a DNA polymerase with 5´→3´ and 3´→5´ exodeoxyribonuclease activities. DNA Polymerase I also incorporates biotinylated nucleotides.•Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
18010017250 U
180100251000 U
Catalog number 18010017
Price (USD)
216.65
Online Exclusive
237.00
Save 20.35 (9%)
Each
Add to cart
Quantity:
250 U
Price (USD)
216.65
Online Exclusive
237.00
Save 20.35 (9%)
Each
Add to cart
DNA Polymerase I is a DNA polymerase with 5´→3´ and 3´→5´ exodeoxyribonuclease activities. DNA Polymerase I also incorporates biotinylated nucleotides.

Applications—DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5´ overhangs
Source—purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Performance and quality testing

Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10 nmol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50 mM potassium phosphate (pH 7.0), 6.7 mM MgCl2 , 1 mM 2-mercaptoethanol, 80 μg/mL template/primer, 32 μM dTTP, 69 nM [3H]dTTP, and enzyme in 100 μL for 30 min at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionDNA Polymerase
Exonuclease Activity3' - 5', 5' - 3'
Hot StartNo
PolymeraseDNA Polymerase I
Quantity250 U
Shipping ConditionWet Ice
FormLiquid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

Is there anything to prevent AmpliTaq Gold DNA polymerase from extending from the 3’ end of a TaqMan probe in a 5’ nuclease assay?

Yes. There is a phosphate group on the 3' end of all TaqMan probes that prevents such extension.

How does AmpliTaq Gold DNA Polymerase differ from AmpliTaq DNA Polymerase?

AmpliTaq Gold DNA Polymerase is a modified form of AmpliTaq DNA Polymerase that contains a proprietary chemical (or so-called hot start molecule) bound to the enzyme's active site. In order to activate the AmpliTaq Gold DNA Polymerase fully, we recommend an initial activation step of 95 degrees C for 10 min when using GeneAmp 10X PCR Buffer I and/or GeneAmp 10X PCR Buffer II and Mg in one of our thermal cyclers. When using GeneAmp 10X PCR Gold Buffer, activation time can be reduced to 5 minutes. Once activation is complete, you can proceed with your standard PCR cycling program (denaturing, annealing, extension, etc).

Does AmpliTaq Gold DNA Polymerase contain exonuclease (proofreading) activity?

No, AmpliTaq Gold DNA polymerase does not contain proofreading activity, however fidelity in PCR amplifications utilizing this enzyme may be improved. High fidelity can be achieved by: 1. Decreasing the final concentration of each nucleotide to 40-50 uM. 2. Using the lowest MgCl2 concentration possible. 3. Using less enzyme. 4. Decreasing extension times. 5. Using the highest annealing temperature possible. 6. Using as few cycles as possible.

Does the fidelity of AmpliTaq DNA Polymerase change in the presence of base analogs?

The fidelity of this PCR enzyme is affected in two ways. First, AmpliTaq DNA Polymerase typically binds to and incorporates base analogs less efficiently than conventional dNTPs, which means that polymerase activity is lower in reactions that contain base analogs. Second, the analog may pair with more than one conventional complementary template base, so the analog may be incorporated at an increased level compared to conventional dNTPs. For the best fidelity, we recommend that base analogs are included at low concentrations in the reaction.

What is the expected half life of AmpliTaq DNA Polymerase at 95 degrees C?

The half-life of AmpliTaq DNA Polymerase at 95 degrees C is 40 min. During PCR, the sample is only incubated at the programmed temperature for approximately 20 seconds. Therefore, the cycling half-life of AmpliTaq Gold at 95 degrees C is approximately 100 cycles.

Example: AmpliTaq DNA Polymerase experiences about 20 seconds at 95 degrees C per PCR cycle. The t1/2 is at least 33 minutes; (35-40 min). Therefore, 33 min/20 sec/cycle = 100 cycles. 100 PCR cycles reduces enzyme activity by 50%.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection