2-Bromobenzyl alcohol, 98%, Thermo Scientific Chemicals
2-Bromobenzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromobenzyl alcohol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number 180100250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18982-54-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point78°C to 82°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only