Butyl chloroformate, 98%, Thermo Scientific Chemicals
Butyl chloroformate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Butyl chloroformate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
Packaging:
Glass bottle
Catalog number 180170010
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
Add to cart
Butyl chloroformate, 98%, Thermo Scientific Chemicals
Catalog number180170010
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS592-34-7
IUPAC Namebutyl carbonochloridate
Molecular FormulaC5H9ClO2
InChI KeyNRDQFWXVTPZZAZ-UHFFFAOYSA-N
SMILESCCCCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<100 APHA
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4110 to 1.4130 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only