Thermo Scientific Chemicals

Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals

Catalog number: 180200010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals

Catalog number: 180200010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 180200010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
91-20-3
IUPAC Name
naphthalene
Molecular Formula
C10H8
InChI Key
UFWIBTONFRDIAS-UHFFFAOYSA-N
SMILES
C1=CC2=CC=CC=C2C=C1
GC
>=99.0 %
Appearance (Form)
Powder or crystals or flakes
Infrared spectrum
Conforms
Solubility
(5 % in methanol) Clear colorless
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For fluorescence/luminescence spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text