Thermo Scientific Chemicals

Silver cyanide, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 180230500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Silver cyanide, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 180230500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 180230500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
506-64-9
IUPAC Name
silver(1+) iminomethanide
Molecular Formula
CAgN
InChI Key
LFAGQMCIGQNPJG-UHFFFAOYSA-N
SMILES
[Ag+].[C-]#N
Iron (Fe)
=<10 ppm
Nickel (Ni)
=<10 ppm
Lead (Pb)
=<10 ppm
Appearance (Color)
White to light grey
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text