Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Catalog number 180301000
Price (USD)/ Each
40.65
Online exclusive
44.50 
Save 3.85 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
44.50 
Save 3.85 (9%)
Add to cart
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000, Thermo Scientific Chemicals
Catalog number180301000
Price (USD)/ Each
40.65
Online exclusive
44.50 
Save 3.85 (9%)
-
Add to cart
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Granular powder
Infrared spectrumConforms
Volatile matter=<5 %
Ash=<1 % (as Na2O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only