T7 RNA Polymerase
T7 RNA Polymerase
Invitrogen™

T7 RNA Polymerase

T7 RNA Polymerase is a DNA-dependent RNA polymerase that has a high specificity for bacteriophage T7 promoter sequences. The enzymeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
180330192500 U
180331002 x 2500 U
Catalog number 18033019
Price (USD)
447.00
Each
-
Add to cart
Quantity:
2500 U
Price (USD)
447.00
Each
Add to cart
Ask our AI about this Product
T7 RNA Polymerase is a DNA-dependent RNA polymerase that has a high specificity for bacteriophage T7 promoter sequences. The enzyme synthesizes large quantities of RNA from DNA inserted into a transcription vector downstream from a T7 promoter. A T7 RNA Polymerase technical bulletin is available.

Application:
Synthesis of labeled and unlabeled RNA transcripts (1).

Source:
Purified from E. coli expressing the T7 RNA Polymerase gene on a plasmid.

Performance and Quality Testing:
3´ and 5´ exodeoxyribonuclease, ribonuclease, and DNA nicking assays; performance in a
transcription reaction.

Unit Definition:
One unit hydrolyzes 1 nmol of ribonucleotide into acid-precipitable material in 1 h at 37°C using a T7 transcription
vector as template.

Unit Reaction Conditions:
40 mM Tris-HCl (pH 8.0), 25 mM NaCl, 8 mM MgCl2 , 2 mM spermidine-(HCl)3 , 5 mM DTT, 0.4 mM ATP, 0.4 mM CTP, 0.4 mM GTP, 0.4 mM UTP, 1 μCi [3H]GTP, 1 μg Sph I-cut pT7L13, and enzyme in 50 μl for 10 min. at 37°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2500 U
Shipping ConditionWet Ice
PolymeraseT7 RNA Polymerase
PromoterT7
Unit SizeEach
Contents & Storage
T7 RNA Polymerase is supplied with a vial of 5X reaction buffer [200 mM Tris-HCl (pH 8.0), 40 mM MgCl2 , 10 mM spermidine-(HCl)3 , 125 mM NaCl], vial of 100 mM DTT. Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3163461Certificate of AnalysisJun 25, 202518033019
3149988Certificate of AnalysisMar 12, 202518033019
3086237Certificate of AnalysisJan 31, 202518033019
2985122Certificate of AnalysisDec 13, 202418033019
2907258Certificate of AnalysisMay 17, 202418033019
5 results displayed, search above for a specific certificate

Safety Data Sheets

Scientific Resources

Product Information

Citations & References (2)

Citations & References
Abstract
Principles of 3' splice site selection and alternative splicing for an unusual group II intron from Bacillus anthracis.
Authors:Robart AR, Montgomery NK, Smith KL, Zimmerly S,
Journal:RNA
PubMed ID:15100440
We investigated the self-splicing properties of two introns from the bacterium Bacillus anthracis. One intron (B.a.I1) splices poorly in vitro despite having typical structural motifs, while the second (B.a.I2) splices well while having apparently degenerated features. The spliced exons of B.a.I2 were sequenced, and splicing was found to occur at ... More
Novel binding of HuR and poly(C)-binding protein to a conserved UC-rich motif within the 3'-untranslated region of the androgen receptor messenger RNA.
Authors: Yeap Bu B; Voon Dominic C; Vivian Julian P; McCulloch Ross K; Thomson Andrew M; Giles Keith M; Czyzyk-Krzeska Maria F; Furneaux Henry; Wilce Matthew C J; Wilce Jackie A; Leedman Peter J;
Journal:J Biol Chem
PubMed ID:12011088
The androgen receptor (AR) mediates androgen action and plays a central role in the proliferation of specific cancer cells. We demonstrated recently that AR mRNA stability is a major determinant of AR gene expression in prostate and breast cancer cells and that androgens differentially regulate AR mRNA decay dependent on ... More
2 total citations

Share catalog number, name or link

1x1 image pixel for data collection