T7 RNA Polymerase
T7 RNA Polymerase
Invitrogen™

T7 RNA Polymerase

T7 RNA Polymerase is a DNA-dependent RNA polymerase that has a high specificity for bacteriophage T7 promoter sequences. The enzymeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
180331002 x 2500 U
180330192500 U
Catalog number 18033100
Price (USD)
730.00
Each
-
Add to cart
Quantity:
2 x 2500 U
Price (USD)
730.00
Each
Add to cart
Ask our AI about this Product
T7 RNA Polymerase is a DNA-dependent RNA polymerase that has a high specificity for bacteriophage T7 promoter sequences. The enzyme synthesizes large quantities of RNA from DNA inserted into a transcription vector downstream from a T7 promoter. A T7 RNA Polymerase technical bulletin is available.

Application:
Synthesis of labeled and unlabeled RNA transcripts (1).

Source:
Purified from E. coli expressing the T7 RNA Polymerase gene on a plasmid.

Performance and Quality Testing:
3´ and 5´ exodeoxyribonuclease, ribonuclease, and DNA nicking assays; performance in a
transcription reaction.

Unit Definition:
One unit hydrolyzes 1 nmol of ribonucleotide into acid-precipitable material in 1 h at 37°C using a T7 transcription
vector as template.

Unit Reaction Conditions:
40 mM Tris-HCl (pH 8.0), 25 mM NaCl, 8 mM MgCl2 , 2 mM spermidine-(HCl)3 , 5 mM DTT, 0.4 mM ATP, 0.4 mM CTP, 0.4 mM GTP, 0.4 mM UTP, 1 μCi [3H]GTP, 1 μg Sph I-cut pT7L13, and enzyme in 50 μl for 10 min. at 37°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 x 2500 U
PolymeraseT7 RNA Polymerase
PromoterT7
Unit SizeEach
Contents & Storage
T7 RNA Polymerase (50 U/μL) is supplied with a vial of 5X reaction buffer [200 mM Tris-HCl (pH 8.0), 40 mM MgCl2 , 10 mM spermidine-(HCl)3 , 125 mM NaCl], vial of 100 mM DTT. Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link