Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Boric acid, 99+%, extra pure
Thermo Scientific Chemicals

Boric acid, 99+%, extra pure

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
1 kg
Packaging:
View all
Plastic bottle
Catalog number 180570010
Price (USD)
58.65
Special offer
Online exclusive
Ends: 26-Sep-2025
72.80
Save 14.15 (19%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 26-Sep-2025
72.80
Save 14.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals or granules
Titration with NaOH99.0 to 101.0 %
Chloride (Cl)=<10 ppm
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link