Thermo Scientific Chemicals

Boric Acid, Powder 99+%, Thermo Scientific Chemicals

Catalog number: 180570100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Boric Acid, Powder 99+%, Thermo Scientific Chemicals

Catalog number: 180570100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 180570100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-35-3
IUPAC Name
boric acid
Molecular Formula
BH3O3
InChI Key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES
OB(O)O
Sulfate (SO4)
=<145 ppm
Titration with NaOH
>=99.0 %
Chloride (Cl)
=<10 ppm
Iron (Fe)
=<10 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text