M-MLV Reverse Transcriptase Buffer
M-MLV Reverse Transcriptase Buffer
Invitrogen™

M-MLV Reverse Transcriptase Buffer

M-MLV Reverse Transcriptase Buffer is for use with Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT).5X buffer composition• 250 mMRead more
Have Questions?
Catalog NumberQuantity
180570181 mL
Catalog number 18057018
Price (EUR)
65,25
Each
-
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (EUR)
65,25
Each
Add to cart
Ask our AI about this Product

M-MLV Reverse Transcriptase Buffer is for use with Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT).

5X buffer composition
• 250 mM Tris-HCl (pH 8.3)
• 375 mM KCl
• 15 mM MgCl2

Included with
• M-MLV Reverse Transcriptase
• SuperScript Reverse Transcriptase
• SuperScript II Reverse Transcriptase
• SuperScript III Reverse Transcriptase

M-MLV Reverse Transcriptase Buffer does NOT contain actinomycin D, nucleotides, or dithiothreitol (DTT).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5X
For Use WithM-MLV Reverse Transcriptase
Quantity1 mL
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Shipping ConditionWet or Dry Ice
Starting MaterialRNA
TechniqueReverse Transcription
Volume1 mL
Unit SizeEach
Contents & Storage

• M-MLV Reverse Transcriptase Buffer (1 mL)
• 0.1 M DTT (500 μL)

Store at –20°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection