Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl terephthalate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 180580010
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.40 
Save 6.75 (14%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.40 
Save 6.75 (14%)
Add to cart
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
Catalog number180580010
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.40 
Save 6.75 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,4-dimethyl benzene-1,4-dicarboxylate
Molecular FormulaC10H10O4
InChI KeyWOZVHXUHUFLZGK-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(=O)OC
Molecular Weight (g/mol)194.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Flakes and/or pellets
Infrared spectrumConforms
Melting point140°C to 143°C
Acid value=<0.03 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only