Lead(IV) acetate, 95%, stabilized, Thermo Scientific Chemicals
Lead(IV) acetate, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(IV) acetate, 95%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number 180620025
Price (USD)/ Each
878.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1033.00 
Save 154.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
878.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1033.00 
Save 154.35 (15%)
Add to cart
Lead(IV) acetate, 95%, stabilized, Thermo Scientific Chemicals
Catalog number180620025
Price (USD)/ Each
878.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1033.00 
Save 154.35 (15%)
-
Add to cart
Chemical Identifiers
CAS546-67-8
IUPAC Nameλ²-lead(2+) tetraacetate
Molecular FormulaC8H12O8Pb
InChI KeyACKFDYCQCBEDNU-UHFFFAOYSA-J
SMILES[Pb++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Iodometric>=94 %
Stabilizerca. 4 to 5 % Acetic acid
Appearance (Color)White to pink-orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only