Myristoyl chloride, 97%, Thermo Scientific Chemicals
Myristoyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Myristoyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 180631000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-64-1
IUPAC Nametetradecanoyl chloride
Molecular FormulaC14H27ClO
InChI KeyLPWCRLGKYWVLHQ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
GC>=96.0 %
Refractive index1.4480 to 1.4500 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only