Thermo Scientific Chemicals

Myristoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 180635000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Myristoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 180635000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 180635000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-64-1
IUPAC Name
tetradecanoyl chloride
Molecular Formula
C14H27ClO
InChI Key
LPWCRLGKYWVLHQ-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC(Cl)=O
Appearance (Color)
Clear colorless to yellow-brown
GC
>=96.0 %
Refractive index
1.4480 to 1.4500 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text