Thermo Scientific Chemicals

Allyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: 180651000
100 mL, Each
Thermo Scientific Chemicals

Allyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: 180651000
100 mL, Each
Quantity
Catalog number: 180651000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
591-87-7
IUPAC Name
prop-2-en-1-yl acetate
Molecular Formula
C5H8O2
InChI Key
FWZUNOYOVVKUNF-UHFFFAOYSA-N
SMILES
CC(=O)OCC=C
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4030 to 1.4050 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text