Thermo Scientific Chemicals

Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals

Catalog number: 180780015
1.5 kg, Each
Thermo Scientific Chemicals

Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals

Catalog number: 180780015
1.5 kg, Each
Quantity
Catalog number: 180780015
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-95-2
IUPAC Name
phenol
Molecular Formula
C6H6O
InChI Key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1
GC
>=99.0 %
Water
=<0.5 %
Residue after evaporation
=<0.05 %
Solidification point
>=40.5°C
Appearance (Color)
Colorless to white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text