Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Thermo Scientific Chemicals

Phenol, 99+%, for biochemistry, loose crystals

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 180785000
Price (USD)
164.65
Special offer
Online exclusive
193.00
Save 28.35 (15%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
164.65
Special offer
Online exclusive
193.00
Save 28.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS108-95-2
IUPAC Namephenol
Molecular FormulaC6H6O
InChI KeyISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Loose crystals or flakes
GC>=99.0 %
Water=<0.5 %
Residue after evaporation=<0.05 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473333Certificate of AnalysisJun 09, 2025180780015, 180781000, 180785000
A0460741Certificate of AnalysisMar 05, 2024180780015, 180781000, 180785000
A0448960Certificate of AnalysisJan 05, 2023180780015, 180781000, 180785000
A0398539Certificate of AnalysisDec 15, 2022180780015, 180781000, 180785000
A0415018Certificate of AnalysisDec 13, 2022180780015, 180781000, 180785000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection